((2R,3S,4R,5R)-5-(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl ((2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)diphosphate Sodium Salt (1:2) 98%

Supplier: Ambeed
Warning

Synonyms: GDP-Mannose disodium salt

A1258789-1MG A1258789-5MG
76680-846EA 174 USD
76680-846 76680-848
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl ((2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)diphosphate Sodium Salt (1:2) 98%
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl ((2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)diphosphate Sodium Salt (1:2)

Precaución: for Research Use Only

Formula: C₁₆H₂₃N₅Na₂O₁₆P₂
MDL Number: MFCD00799981
CAS Number: 148296-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR