5,6-Diaminobenzimidazolinone 98%

Supplier: Ambeed
Warning

A1258697-1G A1258697-250MG A1258697-25G A1258697-5G
76680-838EA 22 USD
76680-838 76680-840 76680-842 76680-844
5,6-Diaminobenzimidazolinone 98%
5,6-Diaminobenzimidazolinone

Precaución: for Research Use Only

Formula: C₇H₈N₄O
MDL Number: MFCD00994340
CAS Number: 55621-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR