1-Chloro-4-(4,4-dimethylcyclohexyl)-2-methylbenzene 97%

Supplier: Ambeed
Warning

A1258663-100G A1258663-1G A1258663-25G
76680-830EA 287 USD
76680-830 76680-832 76680-834
1-Chloro-4-(4,4-dimethylcyclohexyl)-2-methylbenzene 97%
1-Chloro-4-(4,4-dimethylcyclohexyl)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₂₁Cl
MDL Number: MFCD32856364
CAS Number: 2682114-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR