(4S,5R)-4,5-Diphenyl-2-(quinolin-2-yl)-4,5-dihydrooxazole 95%

Supplier: Ambeed
Warning

A1258477-100MG A1258477-1G A1258477-250MG
76680-800EA 50.93 USD
76680-800 76680-802 77621-354
(4S,5R)-4,5-Diphenyl-2-(quinolin-2-yl)-4,5-dihydrooxazole 95%
(4S,5R)-4,5-Diphenyl-2-(quinolin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₄H₁₈N₂O
MW: 350.4125 g/mol
MDL Number: MFCD32858294
CAS Number: 2411386-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR