(SP-4-2)-[4,4′-bis(1,1-dimethylethyl)-2,2′-bipyridine-κN1,κN1′]dibromo-Nickel 97%

Supplier: Ambeed
Danger

A1258470-100MG A1258470-25G A1258470-1G A1258470-5G A1258470-250MG
76680-778EA 24 USD
76680-778 76680-780 77549-494 77549-496 77566-436
(SP-4-2)-[4,4′-bis(1,1-dimethylethyl)-2,2′-bipyridine-κN1,κN1′]dibromo-Nickel 97%
(SP-4-2)-[4,4′-bis(1,1-dimethylethyl)-2,2′-bipyridine-κN1,κN1′]dibromo-Nickel

Precaución: For research use only.

Formula: C₁₈H₂₄Br₂N₂Ni
MDL Number: MFCD32856646
CAS Number: 1894189-67-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR