(2,2'-Bipyridine)nickel dichloride 99%

Supplier: Ambeed
Danger

A1258469-1G A1258469-250MG A1258469-5G A1258469-25G
76680-774EA 21 USD
76680-774 76680-776 77566-434 77596-622
(2,2'-Bipyridine)nickel dichloride 99%
(2,2'-Bipyridine)nickel dichloride

Precaución: For research use only.

Formula: C₁₀H₈Cl₂N₂Ni
MW: 285.780 g/mol
MDL Number: MFCD00614884
CAS Number: 22775-90-2
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR