Bis[2-((4S)-4,5-dihydro-4-isopropyloxazol-2-yl)phenyl]amine 97%

Supplier: Ambeed
Warning

A1258464-100MG A1258464-1G A1258464-250MG
76680-750EA 101.85 USD
76680-750 76680-752 76680-754
Bis[2-((4S)-4,5-dihydro-4-isopropyloxazol-2-yl)phenyl]amine 97%
Bis[2-((4S)-4,5-dihydro-4-isopropyloxazol-2-yl)phenyl]amine

Precaución: for Research Use Only

Formula: C₂₄H₂₉N₃O₂
MW: 391.5100 g/mol
CAS Number: 485394-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR