(11bS)-N-(2,6-Di(naphthalen-1-yl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%

Supplier: Ambeed
Warning

A1258442-100MG A1258442-1G A1258442-250MG A1258442-50MG
76680-720EA 43 USD
76680-720 76680-722 76680-724 76680-726
(11bS)-N-(2,6-Di(naphthalen-1-yl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%
(11bS)-N-(2,6-Di(naphthalen-1-yl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Precaución: for Research Use Only

Formula: C₄₁H₃₃F₃NO₅PS
CAS Number: 2757287-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR