N-((11BS)-2,6-di(naphthalen-2-yl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%

Supplier: Ambeed
Warning

A1258436-100MG A1258436-1G A1258436-50MG A1258436-250MG
76680-714EA 93 USD
76680-714 76680-716 76680-718 77621-348
N-((11BS)-2,6-di(naphthalen-2-yl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%
N-((11BS)-2,6-di(naphthalen-2-yl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Precaución: for Research Use Only

Formula: C₄₁H₂₅F₃NO₅PS
MDL Number: MFCD33021031
CAS Number: 2757287-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR