(4R,4'R,5S,5'S)-2,2'-(Pentane-3,3-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1258434-100MG A1258434-1G A1258434-250MG
76680-702EA 68.96 USD
76680-702 76680-704 76680-706
(4R,4'R,5S,5'S)-2,2'-(Pentane-3,3-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4R,4'R,5S,5'S)-2,2'-(Pentane-3,3-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₃₅H₃₄N₂O₂
MW: 514.6567 g/mol
CAS Number: 1246401-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR