(4S,4'S,5R,5'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1258429-100MG A1258429-1G A1258429-250MG
76680-682EA 90.18 USD
76680-682 76680-684 76680-686
(4S,4'S,5R,5'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) 97%
(4S,4'S,5R,5'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₅₃H₅₄N₂O₂
MW: 751.0081 g/mol
CAS Number: 2361262-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR