(4S,4'S,5R,5'R)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A1258428-250MG
A1258428-100MG
76680-680EA
172.93
USD
76680-680
77621-346
(4S,4'S,5R,5'R)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4S,4'S,5R,5'R)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole)
Precaución: for Research Use Only
Formula:
C₄₅H₃₈N₂O₂ MW: 638.7954 g/mol |
CAS Number:
2361262-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...