(S)-2-(1,3-Bis(4-(tert-butyl)phenyl)-2-((R)-4-isopropyl-4,5-dihydrooxazol-2-yl)propan-2-yl)-4-isopropyl-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1258419-100MG A1258419-1G A1258419-250MG
76680-628EA 132.62 USD
76680-628 76680-630 76680-632
(S)-2-(1,3-Bis(4-(tert-butyl)phenyl)-2-((R)-4-isopropyl-4,5-dihydrooxazol-2-yl)propan-2-yl)-4-isopropyl-4,5-dihydrooxazole 97%
(S)-2-(1,3-Bis(4-(tert-butyl)phenyl)-2-((R)-4-isopropyl-4,5-dihydrooxazol-2-yl)propan-2-yl)-4-isopropyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₃₅H₅₀N₂O₂
MW: 530.7837 g/mol
CAS Number: 2757082-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR