(4S,4'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1258416-100MG A1258416-1G A1258416-250MG
76680-620EA 80.63 USD
76680-620 76680-622 76680-624
(4S,4'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4S,4'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₄₃H₅₀N₂O₂
MW: 626.8693 g/mol
CAS Number: 2172801-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR