(4R,4'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1258413-100MG A1258413-1G A1258413-250MG
76680-608EA 51 USD
76680-608 76680-610 76680-612
(4R,4'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4R,4'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₄₃H₅₀N₂O₂
CAS Number: 2634687-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR