(4R,4'R)-2,2'-(1-Phenylpropane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1258412-100MG A1258412-1G A1258412-250MG
76680-602EA 29 USD
76680-602 76680-604 76680-606
(4R,4'R)-2,2'-(1-Phenylpropane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) 97%
(4R,4'R)-2,2'-(1-Phenylpropane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₉H₃₀N₂O₂
CAS Number: 2634687-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR