(4R,4'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1258409-100MG A1258409-1G A1258409-250MG
76680-596EA 82 USD
76680-596 76680-598 76680-600
(4R,4'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole) 97%
(4R,4'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₄₁H₄₆N₂O₂
CAS Number: 2247206-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR