(4R,4'R)-2,2'-(1-Phenylpropane-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1258408-100MG A1258408-250MG A1258408-1G A1258408-5G
76680-594EA 55 USD
76680-594 77621-342 77867-076 77867-077
(4R,4'R)-2,2'-(1-Phenylpropane-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4R,4'R)-2,2'-(1-Phenylpropane-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₇H₂₆N₂O₂
CAS Number: 2757082-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR