(4R,4'R)-2,2'-(1-Phenylpropane-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1258405-1G A1258405-250MG
76680-584EA 896.47 USD
76680-584 76680-586
(4R,4'R)-2,2'-(1-Phenylpropane-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole) 97%
(4R,4'R)-2,2'-(1-Phenylpropane-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₁H₃₀N₂O₂
MW: 342.4751 g/mol
CAS Number: 2452222-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR