(4R,4'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-methyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1258395-100MG A1258395-1G A1258395-250MG
76680-562EA 150.65 USD
76680-562 76680-564 76680-566
(4R,4'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-methyl-4,5-dihydrooxazole) 97%
(4R,4'R)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-methyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₃₁H₄₂N₂O₂
MW: 474.6774 g/mol
CAS Number: 2828432-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR