(4R,4'R)-2,2'-(Pentane-3,3-diyl)bis(4-methyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1258393-100MG A1258393-1G A1258393-250MG
76680-554EA 150.65 USD
76680-554 76680-556 76680-558
(4R,4'R)-2,2'-(Pentane-3,3-diyl)bis(4-methyl-4,5-dihydrooxazole) 97%
(4R,4'R)-2,2'-(Pentane-3,3-diyl)bis(4-methyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₂O₂
MW: 238.3260 g/mol
CAS Number: 2828432-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR