5-Amino-1-methylquinolin-1-ium iodide 97%

Supplier: Ambeed
Warning

A1258379-250MG A1258379-5G A1258379-1G A1258379-25G
76680-546EA 23 USD
76680-546 76680-548 77596-618 77621-340
5-Amino-1-methylquinolin-1-ium iodide 97%
5-Amino-1-methylquinolin-1-ium iodide

Precaución: for Research Use Only

Formula: C₁₀H₁₁IN₂
MDL Number: MFCD22125886
CAS Number: 42464-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR