(3S,4S)-8-(6-Amino-5-((2-amino-3-chloropyridin-4-yl)thio)pyrazin-2-yl)-3-methyl-2-oxa-8-azaspiro[4.5]decan-4-amine 98%

Supplier: Ambeed
Warning

A1258268-25MG A1258268-250MG A1258268-50MG A1258268-5MG A1258268-100MG
76680-542EA 160 USD
76680-542 77566-432 77621-336 77621-338 77621-334
(3S,4S)-8-(6-Amino-5-((2-amino-3-chloropyridin-4-yl)thio)pyrazin-2-yl)-3-methyl-2-oxa-8-azaspiro[4.5]decan-4-amine 98%
(3S,4S)-8-(6-Amino-5-((2-amino-3-chloropyridin-4-yl)thio)pyrazin-2-yl)-3-methyl-2-oxa-8-azaspiro[4.5]decan-4-amine

Precaución: For research use only.

Formula: C₁₈H₂₄ClN₇OS
MDL Number: MFCD32693919
CAS Number: 1801765-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR