4-Ethylpyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A1258029-100MG A1258029-1G A1258029-250MG
76680-514EA 63 USD
76680-514 76680-516 76680-518
4-Ethylpyridin-2(1H)-one 97%
4-Ethylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₇H₉NO
CAS Number: 37529-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR