trans-Methyl 4-(bromomethyl)cyclohexanecarboxylate 97%

Supplier: Ambeed
Danger

A1257981-1G A1257981-250MG A1257981-5G
76680-510EA 234.46 USD
76680-510 76680-512 77571-544
trans-Methyl 4-(bromomethyl)cyclohexanecarboxylate 97%
trans-Methyl 4-(bromomethyl)cyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₅BrO₂
MW: 235.1182 g/mol
MDL Number: MFCD30290228
CAS Number: 167833-10-5
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR