1-Ethylpyridiniumtetrafluoroborate 98%

Supplier: Ambeed
Danger

A1257767-5G A1257767-25G
76680-508EA 14 USD
76680-508 77867-069
1-Ethylpyridiniumtetrafluoroborate 98%
1-Ethylpyridiniumtetrafluoroborate

Precaución: for Research Use Only

Formula: C₇H₁₀BF₄N
MDL Number: MFCD14156009
CAS Number: 350-48-1
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR