(S)-2-(6-Benzhydrylpyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1257617-100MG A1257617-250MG A1257617-1G
76680-476EA 57 USD
76680-476 76680-478 77867-064
(S)-2-(6-Benzhydrylpyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole ≥97%, ee 99%
(S)-2-(6-Benzhydrylpyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₄H₂₄N₂O
CAS Number: 2417528-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR