(5-Fluoro-2,3-dihydrobenzofuran-4-yl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A1257475-100MG A1257475-250MG A1257475-1G
76680-460EA 29 USD
76680-460 76680-462 77621-310
(5-Fluoro-2,3-dihydrobenzofuran-4-yl)methanamine hydrochloride 97%
(5-Fluoro-2,3-dihydrobenzofuran-4-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁ClFNO
MDL Number: MFCD31714982
CAS Number: 2135600-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR