(R)-N-(4-(tert-Butyl)phenyl)-N-(2-(tert-butylamino)-2-oxo-1-(pyridin-3-yl)ethyl)furan-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: ML188

A1257467-10MG A1257467-1MG A1257467-5MG
76680-454EA 190.97 USD
76680-454 76680-456 76680-458
(R)-N-(4-(tert-Butyl)phenyl)-N-(2-(tert-butylamino)-2-oxo-1-(pyridin-3-yl)ethyl)furan-2-carboxamide 98%
(R)-N-(4-(tert-Butyl)phenyl)-N-(2-(tert-butylamino)-2-oxo-1-(pyridin-3-yl)ethyl)furan-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₆H₃₁N₃O₃
MW: 433.54 g/mol
MDL Number: MFCD32693920
CAS Number: 1417700-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR