2-(2-(Furan-2-yl)phenyl)acetic acid 97%

Supplier: Ambeed
Warning

A1256956-100MG A1256956-1G A1256956-250MG
76680-438EA 40.32 USD
76680-438 76680-440 76680-442
2-(2-(Furan-2-yl)phenyl)acetic acid 97%
2-(2-(Furan-2-yl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃
MW: 202.2060 g/mol
MDL Number: MFCD24500111
CAS Number: 568630-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR