7-Bromo-5-methyl-1,2,3,4-tetrahydroisoquinoline 98%

Supplier: Ambeed
Warning

A1256272-100MG A1256272-250MG A1256272-1G
76680-434EA 185.66 USD
76680-434 76680-436 77596-608
7-Bromo-5-methyl-1,2,3,4-tetrahydroisoquinoline 98%
7-Bromo-5-methyl-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrN
MW: 226.1130 g/mol
MDL Number: MFCD31924026
CAS Number: 1934800-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR