Bis(1,5-cyclooctadiene)rhodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 98%

Supplier: Ambeed
Warning

A1256249-100MG A1256249-1G A1256249-250MG A1256249-50MG A1256249-25MG
76680-426EA 224.92 USD
76680-426 76680-428 76680-430 76680-432 77596-606
Bis(1,5-cyclooctadiene)rhodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 98%
Bis(1,5-cyclooctadiene)rhodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

Precaución: for Research Use Only

Formula: C₄₈H₂₈BF₂₄Rh
MW: 1174.4141 g/mol
MDL Number: MFCD10567026
CAS Number: 404573-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR