Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate Fe 12-14%

Supplier: Ambeed
Warning

Synonyms: Ethylenediaminetetraaceticacidferricsodiumsalttrihydrate, (Ethylenedinitrilo)tetraacetic acid ferric sodium salt, EDTA ferric sodium salt

A1255856-1000G A1255856-100G A1255856-25G A1255856-500G
76680-406EA 132 USD
76680-406 76680-408 76680-410 76680-412
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate Fe 12-14%
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₂FeN₂NaO₈·3H₂O
MW: 421.09 g/mol
MDL Number: MFCD00798117
CAS Number: 18154-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR