2-(2-(Thiophen-3-yl)phenyl)acetic acid 98%

Supplier: Ambeed
Warning

A1255786-1G A1255786-250MG
76680-390EA 239.77 USD
76680-390 76680-392
2-(2-(Thiophen-3-yl)phenyl)acetic acid 98%
2-(2-(Thiophen-3-yl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂S
MW: 218.2716 g/mol
MDL Number: MFCD31380057
CAS Number: 2004388-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR