(3-Hydroxy-4-(methoxycarbonyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A1255567-25G A1255567-250MG A1255567-5G
76680-374EA 978 USD
76680-374 77867-050 77867-051
(3-Hydroxy-4-(methoxycarbonyl)phenyl)boronic acid 98%
(3-Hydroxy-4-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₅
MW: 195.9651 g/mol
MDL Number: MFCD18383893
CAS Number: 1207726-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR