tert-Butyl methyl(5-oxopentyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: 5-[Boc(methyl)amino]pentanal

A1255387-250MG A1255387-100MG A1255387-1G A1255387-50MG
76680-368EA 335.25 USD
76680-368 77621-284 77621-286 77621-288
tert-Butyl methyl(5-oxopentyl)carbamate 97%
tert-Butyl methyl(5-oxopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.2893 g/mol
MDL Number: MFCD30474648
CAS Number: 1620280-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR