6-(Aminomethyl)pyridin-2(1H)-one hydrobromide 97%

Supplier: Ambeed
Warning

A1255361-1G A1255361-250MG
76680-364EA 319 USD
76680-364 76680-366
6-(Aminomethyl)pyridin-2(1H)-one hydrobromide 97%
6-(Aminomethyl)pyridin-2(1H)-one hydrobromide

Precaución: for Research Use Only

Formula: C₆H₉BrN₂O
CAS Number: 95851-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR