3-(5-Bromopyridin-3-yl)phenol 97%

Supplier: Ambeed
Warning

A1252002-1G A1252002-250MG
76680-360EA 140 USD
76680-360 77867-044
3-(5-Bromopyridin-3-yl)phenol 97%
3-(5-Bromopyridin-3-yl)phenol

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO
MDL Number: MFCD25957483
CAS Number: 1276123-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR