Isopropyl (1-((5-chloro-8-hydroxyquinolin-7-yl)methyl)piperidin-4-yl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: ML418

A1251852-10MG A1251852-1MG A1251852-50MG A1251852-5MG
76680-350EA 71.09 USD
76680-350 76680-352 76680-354 76680-356
Isopropyl (1-((5-chloro-8-hydroxyquinolin-7-yl)methyl)piperidin-4-yl)carbamate 98%
Isopropyl (1-((5-chloro-8-hydroxyquinolin-7-yl)methyl)piperidin-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₉H₂₄ClN₃O₃
MW: 377.8652 g/mol
MDL Number: MFCD32201056
CAS Number: 1928763-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR