2-(4-(2-((1H-Benzo[d]imidazol-2-yl)thio)ethyl)piperazin-1-yl)-N-(6-methyl-2,4-bis(methylthio)pyridin-3-yl)acetamide dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: K-604 2HCl

A1251721-10MG A1251721-1MG A1251721-5MG
76680-344EA 114 USD
76680-344 76680-346 76680-348
2-(4-(2-((1H-Benzo[d]imidazol-2-yl)thio)ethyl)piperazin-1-yl)-N-(6-methyl-2,4-bis(methylthio)pyridin-3-yl)acetamide dihydrochloride 98%
2-(4-(2-((1H-Benzo[d]imidazol-2-yl)thio)ethyl)piperazin-1-yl)-N-(6-methyl-2,4-bis(methylthio)pyridin-3-yl)acetamide dihydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₃₂Cl₂N₆OS₃
MW: 575.6408 g/mol
MDL Number: MFCD30738203
CAS Number: 217094-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR