2-Amino-N-(4-aminophenyl)acetamide dihydrochloride 97%

Supplier: Ambeed
Warning

A1251603-100MG A1251603-1G A1251603-250MG
76680-338EA 24 USD
76680-338 76680-340 76680-342
2-Amino-N-(4-aminophenyl)acetamide dihydrochloride 97%
2-Amino-N-(4-aminophenyl)acetamide dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃Cl₂N₃O
MDL Number: MFCD31434089
CAS Number: 2137859-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR