6-Methylpiperidine-2-carboxylic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A1251394-1G
A1251394-250MG
A1251394-5G
76680-318EA
132
USD
76680-318
76680-320
76680-322
6-Methylpiperidine-2-carboxylic acid hydrochloride 97%
6-Methylpiperidine-2-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₄ClNO₂ |
MDL Number:
MFCD16885718 CAS Number: 117928-49-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...