(E)-3-(3,5-Dihydroxyphenyl)acrylic acid 98%

Supplier: Ambeed
Warning

A1251058-100MG A1251058-1G A1251058-250MG
76680-312EA 51.99 USD
76680-312 76680-314 76680-316
(E)-3-(3,5-Dihydroxyphenyl)acrylic acid 98%
(E)-3-(3,5-Dihydroxyphenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.1574 g/mol
MDL Number: MFCD03002833
CAS Number: 127791-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR