2,4,5,6-Tetrakis(diphenylamino)isophthalonitrile 95%

Supplier: Ambeed
Warning

A1250858-100MG A1250858-250MG A1250858-1G A1250858-50MG A1250858-5G A1250858-25G
76680-302EA 58.35 USD
76680-302 76680-304 77549-478 77596-554 77596-556 77621-244
2,4,5,6-Tetrakis(diphenylamino)isophthalonitrile 95%
2,4,5,6-Tetrakis(diphenylamino)isophthalonitrile

Precaución: for Research Use Only

Formula: C₅₆H₄₀N₆
MW: 796.96 g/mol
MDL Number: MFCD32856534
CAS Number: 1846598-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR