Ethyl 3-hydroxy-1-methyl-6-nitro-2-oxo-1,2-dihydroquinoline-4-carboxylate 97%

Supplier: Ambeed
Warning

A1250750-100MG A1250750-250MG A1250750-5G
76680-282EA 26 USD
76680-282 76680-284 76680-286
Ethyl 3-hydroxy-1-methyl-6-nitro-2-oxo-1,2-dihydroquinoline-4-carboxylate 97%
Ethyl 3-hydroxy-1-methyl-6-nitro-2-oxo-1,2-dihydroquinoline-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₆
MW: 292.2442 g/mol
CAS Number: 2230408-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR