(3aR,8aR)-4,4,8,8-Tetrakis(3,5-dimethylphenyl)-N,N,2,2-tetramethyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-amine 97%

Supplier: Ambeed
Warning

A1250744-100MG A1250744-1G A1250744-250MG
76680-280EA 88.06 USD
76680-280 77571-500 77621-240
(3aR,8aR)-4,4,8,8-Tetrakis(3,5-dimethylphenyl)-N,N,2,2-tetramethyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-amine 97%
(3aR,8aR)-4,4,8,8-Tetrakis(3,5-dimethylphenyl)-N,N,2,2-tetramethyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-amine

Precaución: for Research Use Only

Formula: C₄₁H₅₀NO₄P
MW: 651.8138 g/mol
MDL Number: MFCD32691167
CAS Number: 840454-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR