(3aS,6S,7aS)-7,7-Dichloro-8,8-dimethylhexahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide 99%

Supplier: Ambeed
Warning

A1250067-1G A1250067-250MG A1250067-5G
76680-268EA 31.83 USD
76680-268 76680-270 76680-272
(3aS,6S,7aS)-7,7-Dichloro-8,8-dimethylhexahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide 99%
(3aS,6S,7aS)-7,7-Dichloro-8,8-dimethylhexahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide

Precaución: for Research Use Only

Formula: C₁₀H₁₅Cl₂NO₂S
MW: 284.2026 g/mol
CAS Number: 151556-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR