4-Oxo-4-(((2R,3R,4S,5S,6R)-3,4,5-triacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)amino)butanoic acid 97%

Supplier: Ambeed
Warning

A1249841-100MG A1249841-250MG
76680-254EA 22 USD
76680-254 76680-256
4-Oxo-4-(((2R,3R,4S,5S,6R)-3,4,5-triacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)amino)butanoic acid 97%
4-Oxo-4-(((2R,3R,4S,5S,6R)-3,4,5-triacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)amino)butanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₅NO₁₂
CAS Number: 83306-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR