4-(4-Methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline 98%

Supplier: Ambeed
Warning

A1249417-100MG A1249417-1G
76680-244EA 250.38 USD
76680-244 76680-246
4-(4-Methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline 98%
4-(4-Methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline

Precaución: for Research Use Only

Formula: C₁₇H₁₇F₃N₄
MW: 334.3389 g/mol
MDL Number: MFCD00600194
CAS Number: 109028-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR