4-(4-Methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline maleate 98%

Supplier: Ambeed
Warning

Synonyms: CGS-12066 maleate

A1249405-100MG A1249405-1G A1249405-250MG A1249405-50MG
76680-236EA 72 USD
76680-236 76680-238 76680-240 76680-242
4-(4-Methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline maleate 98%
4-(4-Methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline maleate

Precaución: for Research Use Only

Formula: C₂₁H₂₁F₃N₄O₄
MW: 450.4111 g/mol
MDL Number: MFCD01529924
CAS Number: 1350965-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR